Double Cylinder Hand Truck with Firewall and Hoist Ring, Justrite®
Supplier: Justrite
35048 35050 35052
76292-510EA 4252.94 CAD
76292-510 76292-512 76292-514
Double Cylinder Hand Truck with Firewall and Hoist Ring, Justrite®
Trolleys
This Double Cylinder Hand Truck with Firewall and Hoist Ring provides safety when moving and storing of gas cylinders.
  • Steel encased Marinite® I creates a 1 hour fire barrier between the fuel and oxidizer
  • Locking posts and collar plates ensure total cylinder immobilization for hoisting applications
  • Configured with steel wheels with dual ball bearings for easier rolling
  • Rated at 272 kg (600 lbs.) weight capacity
  • Common in welding applications, up to 330 mm (13") and 254 mm (10") in diameter, respectively
  • Constructed using 32 mm (1.25") diameter round steel
More Product Information