Single Cylinder Hand Truck, Justrite®
Supplier: Justrite
35008
76292-472EA 1570.19 CAD
76292-472
Single Cylinder Hand Truck, Justrite®
Trolleys
This Single Cylinder Hand Truck provides safety when moving and storing of gas cylinders.
  • Configured with 254 mm (10") steel hub, foam filled flat-free wheels with dual ball bearings for easier rolling
  • Rated at 454 kg (1000 lbs.) weight capacity
  • Holds one cylinder from 155 to 305 mm (6 to 12") in diameter
  • Provide optimum safety, strength, and longevity
  • Constructed of 25 mm (1") and 32 mm (1.25") diameter steel tube with continuous welding
  • Straps feature a steel cinch buckle and hold individual cylinders securely for safe transport
Due to their weight,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Material Steel
Cylinder type Compressed gas
Strap type Poly Cinch Strap with Steel Buckle (1 per cylinder)