Cylinder Locking Post and Collar Set for Double Cylinder Hand Trucks, Justrite®
Supplier: Justrite
35384
76292-462EA 424.91 CAD
76292-462
Cylinder Locking Post and Collar Set for Double Cylinder Hand Trucks, Justrite®
Trolleys
This Cylinder Locking Post and Collar Set for Double Cylinder Hand Trucks provide safety when moving and storing of gas cylinders.
Set of two locking posts used to lock gas cylinders down securely onto any double cylinder Justrite® hand truck.
For cylinders 7 to 10" in diameter, posts include adjustable ring plates and feature powder coating for added durability.
Commonly used to secure high-pressure cylinders for maximum safety, but also compatible with low-pressure cylinders.