Gas Cylinder Mobile Stands with Locking Post, Justrite®
Supplier: Justrite
35286 35292
76292-394EA 1141.28 CAD
76292-394 76292-400
Gas Cylinder Mobile Stands with Locking Post, Justrite®
Drum Storage
These Gas Cylinder Mobile Stands with Locking Post provide safety when moving and storing of gas cylinders.
  • Locking collar and post system adds vise-like holding capability to immobilize the cylinder
  • Use of safety cylinder caps ensures DOT compliance
  • Exclusive bumper guard edging with steel reinforced vinyl protects cylinders from damage, while providing extra grip
  • Suitable for over-the-road gas cylinder transportation
  • Accommodates cylinders from 4 to 10" (102 to 254 mm) in diameter
  • Locking post extends to 66" (1677 mm) in height
The fully weldedMore Product Information