Cryogenic Gas Cylinder Barricade Rack, Justrite®
Supplier: Justrite
35184 35186 35188 35190
76292-326EA 2012.18 CAD
76292-326 76292-328 76292-330 76292-332
Cryogenic Gas Cylinder Barricade Rack, Justrite®
Furniture Storage Cabinets
This cryogenic gas cylinder barricade rack provides safety when moving and storing of gas cylinders.
  • Secure cryogenic cylinders up to 83.8 cm (33") in diameter
  • For indoor or outdoor cryogenic cylinder storage
  • Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
  • Barricade racks feature two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
Barricade racks surround cylinders with 51 mm (2") square steel structural tubing, continuouslyMore Product Information