Digi-Sense® Temp16 Single-Input RTD Thermometer, Cole-Parmer
Supplier: Antyila Scientific
91428-07
76292-174EA 1069.72 CAD
76292-174
Digi-Sense® Temp16 Single-Input RTD Thermometer, Cole-Parmer
Thermometers Digital Thermometers
Digi-Sense® temp16 single-input RTD thermometer offers versatility and accuracy in a compact design.
  • Three way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Low battery indicator
RTD measurement type.
Ordering information: Probes are sold separately.
Delivery information: Includes protective rubber armor/boot with stand and three AA batteries.