Ao Absorbance Microplate Reader, Azure Biosystems
Supplier: Azure Biosystems
AC3100 AC3101 AC3102 AC3103 AC3104 AC3115 AC3116 AC3105 AC3117 AC3121 AC3122 AC3123 AC3107 AC3108 AC3109 AC3110 AC3124 AC3125 AC3111 AC3126 AC3112 AC3127 AC3113 AC3128 AC3114 AC3201 AC3118 AC3119 AC3120 AC3106
76291-986EA 367.67 CAD
76291-986 76291-988 76291-990 76291-992 76291-994 76291-996 76291-998 76292-000 76292-002 76292-004 76292-006 76292-008 76292-010 76292-012 76292-014 76292-016 76292-018 76292-020 76292-022 76292-024 76292-026 76292-028 76292-030 76292-032 76292-034 76292-038 76292-040 76292-042 76292-044 76292-046
Ao Absorbance Microplate Reader, Azure Biosystems
Microplate Readers
96 well plate absorbance microplate reader.
  • Offers a simple user interface
  • Absorbance-based reading offers maximum sensitivity
  • Best read speeds for kinetic assays
  • High flexibility
  • Lower cost compared to fluorescent and luminescent-based readers
Its top-reading design is ideal for solution-based assays such as ELISAs, DNA and protein quantification, and enzyme kinetics.
Stand-alone, top absorbance reader. Large, easy-to-use 7" touch screen interface. Pre-set and custom protocols. Single and dual wavelength detection modes. 8 position filter wheel: 5More Product Information