Rubber Stopper for Serum Bottles and Vials, Ace Glass Incorporated
Supplier: Ace Glass
5531-47
76291-836CS 443.6 CAD
76291-836
Rubber Stopper for Serum Bottles and Vials, Ace Glass Incorporated
Stoppers Rubber Stoppers
Gray, thin, flange-style chlorobutyl rubber stopper fits all 20 mm serum bottles and serum vials.