Electronic Overhead Stirrers, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
F201A0152 F201A0155 F201A0156 F201A0150 F201A0151 SA201A0151 SA201A0155
76288-116EA 1921.49 CAD
76288-116 76288-118 76288-120 76288-172 76288-174 76288-176 76288-178
Electronic Overhead Stirrers, VELP Scientifica
Stirrers Overhead Stirrers
Complete range of overhead stirrers with electronic adjustment for a variety of needs in terms of viscosity and volume.
  • Wide range of applications: solubilization, mixing, emulsification and homogenization
  • Self-locking chuck simplifies assembly
  • Gentle start-up ensures optimum progression of the stirring speed
  • Most advanced safety standards
  • Through-hole to accept long shafts
Overhead stirrers with a technopolymer structure, ideal for premium resistance to acids, bases and solvents. Many reliable solutions are available, according to differentMore Product Information