Vaplock™ Nalgene® Waste Caps and Waste Kits, 100-415, Cole-Parmer
Supplier: Antyila Scientific
12019-46 12019-47 12019-48 12019-49 12019-50 12019-51
76287-562KT 1025.28 CAD
76287-562 76287-564 76287-566 76287-568 76287-570 76287-572
Vaplock™ Nalgene® Waste Caps and Waste Kits, 100-415, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conformable to a broad range of containment types
Featuring a vapor tight connection, waste manifolds are designed to fit Nalgene® 100-415 lab bottles. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable