Cryogenic Thermal Transfer Labels for Frozen Vials and Tubes
Supplier: GA INTERNATIONAL INC
807656026862 807656026879 807656026886 807656026893
76276-190RL 225.62 CAD
76276-190 76276-192 76276-194 76276-196
Cryogenic Thermal Transfer Labels for Frozen Vials and Tubes
Labels
These labels are intended for identifying already frozen vials and tubes under deep-freeze conditions (−80 °C) prior to storage in ultra-low temperature lab freezers and liquid nitrogen tanks (−196 °C).
  • Temperature range: −196 to 110 °C (−321 to 230 °F)
  • Labels can adhere to already frozen vials (no thawing required)
  • Ideal for long-term storage in cryogenic conditions (−196 °C)
  • Ideal for safely thawing in water baths
  • Printable with all major thermal-transfer printer brands
An ideal solution for bio-banks and bio-repositories that need labeling orMore Product Information