DYMO Compatible Cryogenic Labels
Supplier: GA INTERNATIONAL INC
807656032917 807656032788 807656032771 807656032726 807656032696 807656032719 807656032702 807656032818 807656032870 807656032856 807656032849 807656032832 807656032863 807656032764 807656032757 807656032740 807656032733 807656032689 807656032795 807656032900 807656032887 807656032801 807656032825 807656032894 807656024370 807656024387 807656024394 807656024400 807656024332 807656024349 807656024356 807656024363 807656024219 807656006864 807656005898 807656024226 807656024233 807656024240 807656006512 807656024257 807656005232 807656024264 807656024271 807656005461 807656024288 807656024295 807656024301 807656005355 807656024196 807656024202 807656005348 807656024325 807656024417 807656024424 807656024431 807656024455 807656024462 807656024479 807656024486 807656024493 807656024509 807656024516 807656024523 807656024530 807656024547 807656024554 807656024561 807656005324 807656006642 807656006635 807656006628 807656006611 807656006277 807656032924
76210-294RL 132.04 CAD
76210-294 76210-300 76210-302 76210-304 76210-306 76210-308 76210-310 76210-312 76210-314 76210-316 76210-318 76210-320 76210-322 76210-490 76210-492 76210-494 76210-496 76210-594 76210-596 76210-598 76210-600 76210-602 76210-604 76210-606 76274-294 76274-296 76274-298 76274-300 76274-654 76274-656 76274-658 76274-660 76275-286 76275-288 76275-290 76275-292 76275-294 76275-296 76275-298 76275-300 76275-302 76275-304 76275-306 76275-308 76275-310 76275-312 76275-314 76275-320 76275-322 76275-324 76275-402 76275-404 76275-406 76275-408 76275-410 76275-414 76275-416 76275-418 76275-420 76275-422 76275-424 76275-426 76275-428 76275-430 76275-432 76275-434 76275-436 76275-438 76275-440 76275-442 76275-444 76275-454 76275-456 77321-180
DYMO Compatible Cryogenic Labels
Labels
DYMO compatible direct thermal cryo labels with permanent adhesive.
  • Temperature range: –196 to +70 °C (–321 to +158 °F)
  • Compatible with all DYMO label writers (excluding models 550, 550Turbo, and 5XL)
  • Resists extreme cryogenic temperatures (–196° C in liquid phase and vapour phase liquid nitrogen)
  • Ideal for the identification of 0.5 to 5 ml vials and tubes
DYMO compatible cryo labels are available in a range of sizes, shapes, and colors, perfect for identifying 0.5 to 5 ml cryo vials and microtubes, vial caps/tops, cryo/freezer boxes, as well asMore Product Information