Cryogenic Labels for Inkjet Printers
Supplier: GA INTERNATIONAL INC
807657630631
76272-006PK 140.48 CAD
76272-006 76272-008 76273-220 76273-222 76273-226 76274-842 76274-844 76274-846 76274-848 76274-906 76274-912 76274-914 76274-916 76274-920 76274-922 76274-924 76274-958 76274-960
Cryogenic Labels for Inkjet Printers
Labels
Cryogenic inkjet labels, provided in a roll format for printing with roll format inkjet printers. These cryo labels withstand prolonged storage in cryogenic conditions, including liquid nitrogen (−196 °C) and laboratory freezers (−80 °C), as well as wiping and rubbing with alcohol sanitizing wipes.
  • Temperature range: −196 to +121 °C (−321 to +250 °F)
  • Resist extreme cryogenic (−196 °C, liquid/vapor phase liquid nitrogen) and deep-freeze temperatures (−80 °C, low temperature freezers)
  • Print eye-catching color logos, graphics, images, and data
  • Resists wiping
More Product Information