Wrap-Around Cryogenic Thermal Transfer Labels
Supplier: GA INTERNATIONAL INC
807656033419 807656033426 807656017402 807656017419 807656017426 807656017358 807656027883 807656017501 807656017518 807656017525 807656017327 807656017334 807656017433 807656017440 807656017457 807656017464 807656017471 807656017488 807656017495 807656027876 807656017532 807656017549 807656017556 807656017563 807656028491 807656017570 807656017587 807656017594 807656017600 807656017617 807656017624 807656017631 807656017648 807656028507 807656017655 807656017662 807656017679 807656017686 807656017693 807656017709 807656017716 807656017723 807656017730 807656017747 807656017754 807656017761 807656017778
76210-074RL 169.72 CAD
76210-074 76210-076 76273-344 76273-346 76273-348 76273-510 76273-918 76273-920 76273-922 76273-924 76273-976 76273-978 76273-982 76273-984 76273-986 76273-988 76273-990 76273-992 76273-994 76273-996 76273-998 76274-000 76274-002 76274-004 76274-006 76274-008 76274-010 76274-012 76274-014 76274-016 76274-018 76274-020 76274-022 76274-024 76274-026 76274-028 76274-030 76274-032 76274-034 76274-036 76274-038 76274-040 76274-042 76274-044 76274-046 76274-048 76274-050
Wrap-Around Cryogenic Thermal Transfer Labels
Labels
Transparent cryogenic self-laminating labels for thermal transfer printers. Their clear laminate protects the printed information from alcohols and detergents to ensure the secure identification of tubes and cryo vials.
  • Temperature range: −196 to +100 °C (−321 to +212 °F)
  • Resist cryogenic conditions storage such as vapor phase and liquid phase liquid nitrogen
  • Labels are self-laminating, protecting the information from abrasion and scratching
  • Able to withstand exposure to harsh chemicals and solvents
  • Permanent adhesion, especially on curved surfaces (cryo
More Product Information