Supplier: GA INTERNATIONAL INC
807656031132 807656031149 807656016276 807656016283 807656016290 807656016306 807656016313 807656016320 807656016337 807656016344 807656015958 807656015965 807656015972 807656027623 807656015989 807656015996 807656016009 807656016016 807656016023 807656016030 807656016047 807656016054 807656016061 807656016078 807656016085 807656016092 807656016108 807656016115 807656016122 807656016139 807656016146 807656016153 807656016160 807656016177 807656016184 807656016191 807656016207 807656016214 807656016221 807656016238 807656016245 807656001562 807656118154 807656118161 807656016351 807656016368 807656016375 807656016382 807656016399 807656016405 807656001548 807656001555 807656016412 807656016429 807656016436 807656016443 807656016450 807656016467 807656016474 807656016481 807656016498 807656016504 807656016511 807656016528 807656016535 807656016559
76210-466RL 113.37 CAD
76210-466 76210-468 76273-646 76273-648 76273-650 76273-652 76273-654 76273-656 76273-658 76273-660 76273-712 76273-714 76273-716 76273-718 76273-720 76273-722 76273-724 76273-726 76273-728 76273-730 76273-732 76273-734 76273-736 76273-738 76273-740 76273-742 76273-744 76273-746 76273-748 76273-750 76273-752 76273-754 76273-756 76273-758 76273-760 76273-762 76273-764 76273-766 76273-768 76273-770 76273-772 76273-774 76273-776 76273-780 76273-782 76273-784 76273-786 76273-788 76273-790 76273-792 76273-794 76273-796 76273-798 76273-800 76273-802 76273-804 76273-806 76273-808 76273-810 76273-812 76273-814 76273-816 76273-818 76273-820 76273-822 76273-824
Labels
Removable deep freeze labels for vials, tubes, and freezer boxes. These thermal transfer freezer labels have a glove-friendly adhesive that ensures the labels can be easily removed without leaving any adhesive residue.
  • Temperature range: –80 to 100 ℃ (–112 to 212 ℉)
  • Ideal for long-term storage in deep-freeze conditions (–80 ℃)
  • Removable adhesive ensures the labels can be easily removed without leaving any adhesive residue
  • Printable with all major thermal-transfer printer brands
  • Resistant to gamma irradiation (tested up to 50 KGy)
Deep freeze,More Product Information