BioClean-D™ Sterile Drop-down Garments, Ansell
Supplier: Ansell Canada
S-BDSH-L S-BDSH-M S-BDSH-S S-BDSH-XL S-BDSH-XS S-BDSH-XXL S-BDSH-XXXL S-BDSH-XXXXL S-BDSH-L S-BDSH-M S-BDSH-S S-BDSH-XL S-BDSH-XS S-BDSH-XXL S-BDSH-XXXL S-BDSH-XXXXL S-BDSH-8XL S-BDSH-5XL S-BDSH-6XL S-BDSH-7XL
75845-792CS 918.74 CAD
75845-792 75845-794 75845-796 75845-798 75845-800 75845-802 75845-804 75845-806 CA75845-792 CA75845-794 CA75845-796 CA75845-798 CA75845-800 CA75845-802 CA75845-804 CA75845-806 76271-366 76271-424 76271-426 76271-428
BioClean-D™ Sterile Drop-down Garments, Ansell
Disposable Garments
S-BDSH with hood is a sterile antistatic disposable garment manufactured from low-linting CleanTough™ material.
  • Elasticated back, hood, cuffs and ankles
  • Foot-loop to aid smooth zip closure
  • Thumb loops ensure a secure hold
  • Quick release press stud tabs for aseptic donning
  • Low-linting CleanTough™ material
The BioClean-D™ drop-down garment's unique design offers true aseptic donning, with internal colored tabs to indicate safe touch points that prevent contact with the outside surface. The innovative up and over donning design eliminates the risk of theMore Product Information