Corepoint Scientific™ White Diamond Series Undercounter and Countertop Refrigerators and Freezers, Horizon Scientific
Supplier: Horizon Scientific
PR051WWW/0 PR051WWWLH/0 PR051SSS/0 PR051SSSLH/0 PR051WWG/0 PR051WWGLH/0 PR051WWGADA/0 PR051WWGADALH0 PF051WWW/0M PF051WWWLH/0M PF051WWWADA/0M PF051WWWADAL0M PF051SSS/0M PF051SSSLH/0M PF051WWW/0A PF051WWWLH/0A PR011WWW/0 PR011WWWLH/0 PR031WWW/0 PR011WWG/0 PR011WWGLH/0 PR031WWG/0 PR061WWG/0 PF021WWW/0M PF021WWWLH/0M PF021WWW/0A PF021WWWLH/0A PF051WWW/0M30 PF051WWW/0M40 FF051WWW/0M FR051WWW/0 PR061WWW/0 PR051WWWADA/0 PR051WWWADALH0
76271-286EA 3320.92 CAD
76271-286 76271-288 76271-290 76271-292 76271-294 76271-296 76271-298 76271-300 76271-302 76271-304 76271-306 76271-308 76271-310 76271-312 76271-314 76271-316 76271-318 76271-320 76271-322 76271-324 76271-326 76271-328 76271-330 76271-332 76271-334 76271-336 76271-338 76271-340 76271-342 76271-344 76271-346 76271-352 76271-354 76271-356
Corepoint Scientific™ White Diamond Series Undercounter and Countertop Refrigerators and Freezers, Horizon Scientific
Refrigerators
These undercounter and countertop refrigerators and freezers provide premium laboratory cold storage for scientific and medical use
  • Integrated temperature alarms
  • Precise microprocessor temperature control
  • Forced draft circulation
  • Keyed door locks
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Select models offer auto defrost cycles, ADA compliance, and leftMore Product Information