Embedding Cassette/Block Storage Cabinet, Avantik
Supplier: Avantik
ST7059 ST7065
76270-344EA 599.85 CAD
76270-344 76270-346
Embedding Cassette/Block Storage Cabinet, Avantik
Furniture Storage Cabinets
Built with proprietary designed drawer technology and interlocking cabinet stackability. The thoughtful design provides more clearance between the tops of the blocks and the drawers, allowing users to open and close them seamlessly.
  • Capacity of up to 1500 cassettes
  • Convenient optional caster wheels allow for easy mobility of the entire cabinet stack
Avantik’s no-jam, hassle-free block storage cabinet is the industry’s first jam-free, hassle-free block storage cabinet. Built with proprietary designed drawer technology and interlocking cabinetMore Product Information