gBrite™ LED Plant Growth Chambers, Extra Large Capacity, Caron Products
Supplier: Caron Products
7310-50-2 7310-50-3 7310-75-2 7310-75-3 7311-50-2 7311-50-3 7311-75-2 7311-75-3 7312-50-2 7312-50-3 7312-75-2 7312-75-3 7317-50-2 7317-50-3 7317-75-2 7317-75-3 7314-50-2 7314-50-3 7314-75-2 7314-75-3
76269-576EA 94996.32 CAD
76269-576 76269-578 76269-580 76269-582 76269-590 76269-592 76269-594 76269-596 76269-604 76269-606 76269-608 76269-610 76269-646 76269-648 76269-650 76269-652 76468-152 76468-154 76468-156 76468-158
gBrite™ LED Plant Growth Chambers, Extra Large Capacity, Caron Products
Testing Chambers Plant Growth Chambers
The new gBrite™ LED-powered Plant Growth Chambers - now Nature has competition.
  • Powerful lighting
  • Easy to maintain and use
  • Facility-friendly design
The gBrite™ LED plant growth chambers incorporate powerful new lighting technology, providing a choice between standard full-spectrum white and optional four-color LED options. LED efficiency reduces plant heat stress, while producing similar light quality to fluorescent, but is unaffected by changes in temperature setpoint. Colored blue/white/red/far-red LED provides a powerful new option, varying bothMore Product Information