Complete Package Melting Point M-565, Buchi
Supplier: Buchi
11060700
76266-270EA 26820.74 CAD
76266-270
Complete Package Melting Point M-565, Buchi
Melting Point Apparatus
Complete Package Melting point M-565, Sample Loader M-569 and software MeltingPoint Monitor.
  • Automated melting, boiling and slip point
  • Video verification and recording
  • Suited for regulated environments
  • Fail-safe calibration
The Melting Point M-565 for the automatic determination of melting and boiling points is designed for true, professional quality control. It includes a color display to observe the phase transition, is equipped with a video camera and a video replay function which allows reviewing the melting and boiling point determination.