xTIP4™ CleanPak™ Pipette Tip Reload, Rainin® LTS® Pipette Compatible, Biotix
Supplier: Biotix
63300010 63300011 63300012 63300013 63300014 63300025 63300026 63300027 63300028 63300029
76266-108CS 394.35 CAD
76266-108 76266-110 76266-112 76266-114 76266-116 76266-118 76266-120 76266-122 76266-124 76266-126
xTIP4™ CleanPak™ Pipette Tip Reload, Rainin® LTS® Pipette Compatible, Biotix
Pipette Tips
Create a perfect ergonomic system in the lab with FlexFit®, StarStop®, and a Biotix xPIPETTE or Rainin LTS® pipette.
  • Low cost option to get high-quality tips
  • 960 tips on the lab bench using the same surface area as 1 rack
  • Re-sealable bag is available in both sterile and nonsterile formats
  • Color-coded snap cards allow for quick identification
  • The product is fully recyclable and contains no contaminating cardboard or paper
  • Inserts are easy to transfer to an Optimus empty rack
  • Full multi-channel and single-channel xPIPETTE and LTS® pipette compatibility