xTIP4™ Racked Pipette Tips, Rainin® LTS® Pipette Compatible, Biotix
Supplier: Biotix
63300007 63300008 63300009 63300020 63300021 63300022 63300023 63300024 63300001 63300002 63300003 63300004 63300005 63300006 63300931
76266-092CS 546.99 CAD
76266-092 76266-094 76266-096 76266-098 76266-100 76266-102 76266-104 76266-106 76266-142 76266-144 76266-146 76266-148 76266-150 76266-152 76314-472
xTIP4™ Racked Pipette Tips, Rainin® LTS® Pipette Compatible, Biotix
Pipette Tips
Biotix xTIP4 pipette tips are fully compatible with Rainin LTS single, multi, and electronic pipettes. Create a perfect ergonomic system in your lab with FlexFit®, StarStop®, and a Biotix xPIPETTE or Rainin LTS® pipette.
  • Polyethylene filters protect against aerosol and liquid contamination with no colorant or CMC contamination
  • Full multi-channel and single-channel LTS and xPIPETTE compatibility
Light Insertion Force
FlexFit® sealing technology requires minimal insertion force to generate consistent seals that users can trust with every tip.