UniStore™ Reagent Bottles, Narrow Mouth, PP
Supplier: United Scientific Supplies
33253 33254 33301 33302 33303 33304 33305
76263-130PK 12.08 CAD
76263-130 76263-132 76263-134 76263-136 76263-138 76263-140 76263-142
UniStore™ Reagent Bottles, Narrow Mouth, PP
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Rigid
  • Autoclavable
UniStore™ narrow mouth PP reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.