Anti-Rorc Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GR8
76263-046EA 834.24 CAD
76263-046
Anti-Rorc Rabbit Polyclonal Antibody
Antibodies
Anti-Nuclear receptor ROR gamma pS203 was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Nuclear receptor ROR gamma pS203 antibody detects mouse receptor ROR gamma phosphorylated at the serine 203 position. RAR-related orphan receptor gamma is a member of the nuclear receptor family of transcription factors. (ROR-gamma) is a key regulator of cellular differentiation, immunity, peripheral circadian rhythm as well as lipid, steroid, xenobiotics and glucose metabolism. ROR-gamma regulates the circadian expression of clock genes such as CRY1, ARNTL/More Product Information
Order Now

SPECIFICATIONS

Antigen symbol Rorc
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Modification Phosphorylation
Purification Affinity purified
Storage temperature Store vial at –20 °C. This product is stable at 4 °C as an undiluted liquid. For extended storage, aliquot contents and freeze at –20 °C or below. Dilute only prior to immediate use.
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR