Anti-COL1A1 Rabbit Polyclonal Antibody (FITC)
Supplier: Rockland Immunochemical
600-402-103
76262-766EA 870.58 CAD
76262-766
Anti-COL1A1 Rabbit Polyclonal Antibody (FITC)
Antibodies
COLLAGEN Type I Fluorescein Conjugated Antibody is specific for Collagen Type I. Collagen Type I (Type-I collagen) is the most abundant collagen of the human body. It is present in scar tissue, the end product when tissue heals by repair. It is also found in tendons, the endomysium of myofibrils and the organic part of bone. Anti-collagen Type I antibody is suitable for Cancer research and other general research.
Anti-COLLAGEN Type I Fluorescein Conjugated Antibody is suitable for western blot, immunoprecipitatioin, Flow Cytometry, and immunohistochemistry. Researchers should determineMore Product Information