Anti-PKM Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GV5S
76262-732EA 229.1 CAD
76262-732
Anti-PKM Rabbit Polyclonal Antibody
Antibodies
Anti-PKM2 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Pyruvate kinase has been shown to be expressed in tumor, embryonic and normal proliferating cells in varying levels. Studies show that when PKM2 is secreted it can induce EGFR phosphorylation which stimulates the EGFR downstream pathway. Previously PKM has been shown to be critical for aerobic glycolysis. Four isoforms of PKM play alternative roles within the system and post-translational modifications along these isoforms induce multiple pathways making this Anti-PKM2More Product Information
Order Now

SPECIFICATIONS

Antigen symbol PKM
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Purification Affinity purified
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR