Anti-MAPK3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GP0S
76262-682EA 229.1 CAD
76262-682
Anti-MAPK3 Rabbit Polyclonal Antibody
Antibodies
ERK1 antibodies detect the ERK1 isoform. Mitogen activiated protein kinase 3, also known as MAPK3, ERK, or ERK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. Anti-ERK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-ERK 1 (RABBIT)More Product Information
Order Now

SPECIFICATIONS

Antigen symbol MAPK3
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Purification Affinity purified
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR