Anti-MAP2K2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GN8
76262-676EA 834.24 CAD
76262-676
Anti-MAP2K2 Rabbit Polyclonal Antibody
Antibodies
MEK2 antibodies detect the MEK2 isoform. Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol MAP2K2
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Purification Affinity purified
Storage temperature Store vial at –20 °C. This product is stable at 4 °C as an undiluted liquid. For extended storage, aliquot contents and freeze at –20 °C or below. Dilute only prior to immediate use.
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR