Hinged Conductive Boxes, All-Spec Industries
Supplier: HISCO, INC TE
665-752 663-111
76248-674EA 56.85 CAD
76248-674 76252-576
Hinged Conductive Boxes, All-Spec Industries
Boxes Storage Boxes
These 660 series conductive plastic boxes are made of a surface and volume conductive materials and it also a volume conductive and electrostatic shielding material.
Certifications: Accordance to EIA standard 514, ASTM D257-75 and EOS/ESD-DS-11.11.1991 test methods.