CHO-FOIL® Shielding Foil and Fabric Tape with Conductive Adhesive, Parker Chomerics
Supplier: PARKER CHOMERICS TE
CCK-36-101-005
76240-672EA 654.14 CAD
76240-672
CHO-FOIL® Shielding Foil and Fabric Tape with Conductive Adhesive, Parker Chomerics
Tapes
Parker Chomerics CHO-FOIL® metal foil tapes provide an economical solution to applications requiring excellent electrical conductivity across substrates. It can be used to wrap cables in order to provide a low impedance connection between cables and backshells. Metal shielding tape is idea for seams of EMI test chambers and enclosures to provide electrical continuity and enhanced shielding.
  • Metal tapes are available in copper, aluminum, and tin-plated copper
  • Embossment available on several metal tape options
  • Tapes available with non-conductive PSA
  • Standard
More Product Information