Human NRCAM ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0851
76237-650EA 1150.24 CAD
76237-650
Human NRCAM ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human NRCAM AccuSignal ELISA Kit
  • Natural and recombinant human NRCAM.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L30-N600
Neuronal cell adhesion molecule is a protein that in humans is encoded by the NRCAM gene. Cell adhesion molecules (CAMs) are members of the immunoglobulin superfamily. This gene encodes a neuronal cell adhesion molecule with multiple immunoglobulin-like C2-type domains and fibronectin type-III domains. This ankyrin-binding protein is involved in neuron-neuron adhesion and promotes directional signaling during axonal cone growth. Also, this gene is expressed in non-neural tissues and may play a general role in cell-cell communication via signaling from its intracellular domain to the actin cytoskeleton during directional cell migration. Allelic variants of this gene have been associated with autism and addiction vulnerability. Alternative splicing results in multiple transcript variants encoding different isoforms.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human NRCAM standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human NRCAM standard solution and each sample be measured in duplicate.
Order Now
SPECIFICATIONS
Environmentally Preferable
Primary antibody reactivity Human
Host
Shipping temperature Wet Ice