Human Livin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0812
76237-500EA 1150.24 CAD
76237-500
Human Livin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Livin AccuSignal ELISA Kit
  • Natural and recombinant human Livin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: M1-S298
BIRC7, also called KIAP or Livin, is a protein that in humans is encoded by the BIRC7 gene. This gene is mapped to 20q13.33. It encodes a member of the inhibitor of apoptosis protein (IAP) family, and contains a single copy of a baculovirus IAP repeat (BIR) as well as a RING-type zinc finger domain. The BIR domain is essential for inhibitory activity and interacts with caspases, while the RING finger domain sometimes enhances antiapoptotic activity but does not inhibit apoptosis alone. Elevated levels of the encoded protein may be associated with cancer progression and play a role in chemotherapy sensitivity. BIRC7 blocked apoptosis induced by BAX induction of cytochrome C release from mitochondria, and it also can block apoptosis induced by chemotherapeutic agents.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human Livin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, cell lysates or tissue homogenates to each empty well. It is recommended that each human Livin standard solution and each sample be measured in duplicate.