Rat ADAM12 ELISA Kit, Rockland Immunochemicals

Supplier: Rockland Immunochemical

KOA0767
76237-474EA 1150.24 CAD
76237-474
Rat ADAM12 ELISA Kit, Rockland Immunochemicals
Assays ELISAs

Rat ADAM12 AccuSignal ELISA Kit


  • Natural and recombinant rat ADAM12.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: R29-S513


ADAM12(Disintegrin and metalloproteinase domain-containing protein 12), also known as MLTN, is an enzyme that in humans is encoded by the ADAM12 gene. This gene encodes a member of the ADAM (a disintegrin and metalloprotease) protein family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene has two alternatively spliced transcripts: a shorter secreted form and a longer membrane-bound form. The shorter form is found to stimulate myogenesis. By RT-PCR and immunoblot analyses that expression of mouse Adam12 increases during muscle regeneration, while the levels of other ADAMs remain constant. Immunofluorescence analysis revealed staining of small, newly formed muscle fibers in regenerating but not normal adult muscle cells. By using of fluorescence in situ hybridization, the ADAM12 gene is mapped to human chromosome 10q26.3.


Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml rat ADAM12 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each rat ADAM12 standard solution and each sample be measured in duplicate.

Order Now

SPECIFICATIONS

Environmentally Preferable
Primary antibody reactivity
Assay Type Sandwich
Host
Shipping temperature Wet Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR