Human IL-31 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0607
76237-444EA 1150.24 CAD
76237-444
Human IL-31 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-31 AccuSignal ELISA Kit
  • Natural and recombinant human IL-31.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S24-T164
Interleukin-31, also known as IL-31 is a protein that in humans is encoded by the IL31 gene. This gene is mapped to 12q24.31. IL31, which is made principally by activated Th2-type T cells, interacts with a heterodimeric receptor consisting of IL31RA and OSMR that is constitutively expressed on epithelial cells and keratinocytes. IL31 may be involved in the promotion of allergic skin disorders and in regulating other allergic diseases, such as asthma. IL31 may function in skin immunity.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human IL-31 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-31 standard solution and each sample be measured in duplicate.