Mouse CXCL13 - BLC ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0433
76237-292EA 1150.24 CAD
76237-292
Mouse CXCL13 - BLC ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CXCL13 - BLC AccuSignal ELISA Kit
  • Natural and recombinant mouse CXCL13.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: I22-A109
C-X-C motif chemokine 13(CXCL13), also known as B lymphocyte chemoattractant(BLC), is a protein that in humans is encoded by theCXCL13 gene. CXCL13 is a small cytokine belonging to the CXC chemokine family, and it was located on sequence maps to 4q21, near the genes encoding most CXC chemokines. In T-lymphocytes, CXCL13 expression is thought to reflect a germinal center origin of the T-cell. Hence, expression of CXCL13 in T-cell lymphomas, such as Angioimmunoblastic T-cell Lymphoma, is thought to reflect a germinal center origin of the neoplastic T-cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse CXCL13 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CXCL13 standard solution and each sample be measured in duplicate.