Human Cathepsin B ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0385
76237-236EA 1110.74 CAD
76237-236
Human Cathepsin B ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Cathepsin B AccuSignal ELISA Kit
  • Natural and recombinant human Cathepsin B.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: R18-I339
Cathepsin B is an enzymatic protein belonging to the peptidase or protease families. In humans, it is coded by the CTSB gene.1, 2 And this gene is mapped to chromosome 8p22.3 The protein encoded by this gene is a lysosomal cysteine proteinase composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. It is a member of the peptidase C1 family. Cathepsin B was once suspected as a candidate protease participating in the conversion of beta-amyloid precursor protein into the amyloid plaques found in Alzheimer's disease patients. However, this function is now known to be mediated by BACE1 protease. It is now thought that cathepsin B can degrade beta-amyloid precursor protein into harmless fragments. Thus, it is conceivable cathepsin B may play a pivotal role in the natural defense against Alzheimer's disease.4 Overexpression of cathepsin B has been associated with esophageal adenocarcinoma and other tumors. At least five transcript variants encoding the same protein have been found for this gene. The standard product used in this kit is recombinant human Cathepsin B with the molecular mass of 37KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Cathepsin B standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Cathepsin B standard solution and each sample be measured in duplicate.