Mouse Oncostatin M - OSM ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0284
76236-764EA 1150.24 CAD
76236-764
Mouse Oncostatin M - OSM ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Oncostatin M - OSM AccuSignal ELISA Kit
  • Natural and recombinant mouse Oncostatin M.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A24-R206
OSM(Oncostatin M) is a protein that in humans is encoded by the OSM gene. OSM is a pleiotropic cytokine that belongs to the interleukin 6 group of cytokines. The OSM gene is mapped on 22q12.2. Of these cytokines it most closely resembles leukemia inhibitory factor(LIF) in both structure and function. Treatment with recombinant OSM leads to the inhibition of proliferation and changes in cellular morphology of a number of tumor cell lines derived from a wide variety of tissue types. OSM signals through cell surface receptors that contain the protein gp130. The type I receptor is composed of gp130 and LIFR, the type II receptor is composed of gp130 and OSMR.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse Oncostatin M standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Oncostatin M standard solution and each sample be measured in duplicate.