Human IL-4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0220
76236-664EA 1150.24 CAD
76236-664
Human IL-4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-4 AccuSignal ELISA Kit
  • Natural and recombinant human IL-4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: H25-S153
Interleukin-4(IL-4), also knowns as a B-cell stimulatory factor1(BSF1), is an immunomodulatory cytokine, which can inhibit the growth of tumour cells.The human cDNA contains a single open reading frame encoding a protein of 153 amino acids, including a putative signal peptide. IL-4 may act as an autocrine growth factor in pancreatic cancer cells and also give rise to the possibility that cancer-derived IL-4 may suppress cancer-directed immunosurveillance in vivo in addition to its growth-promoting effects, thereby facilitating pancreatic tumor growth and metastasis. The mouse and human genes and their protein products show structural and functional similarities. The human IL-4 gene, which occurs as a single copy in the haploid genome, is mapped on chromosome 5. The standard product used in this kit is recombinant human IL-4, consisting of 130 amino acids with the molecular mass of 14KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human IL-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-4 standard solution and each sample be measured in duplicate.