Rat IL-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0216
76236-656EA 1150.24 CAD
76236-656
Rat IL-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat Interleukin-2 AccuSignal ELISA Kit
  • Natural and recombinant rat IL-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A21-Q155
Interleukin-2(IL2), formerly referred to as T-cell growth factor, is a powerfully immunoregulatory lymphokine that is produced by lectin- or antigen-activated T cells. It is produced not only by mature T lymphocytes on stimulation but also constitutively by certain T-cell lymphoma cell lines. The lymphokine interleukin-2(IL-2) is responsible for autocrine cell cycle progression and regulation of immune responses. IL-2 expression in mature thymocytes and T cells has been found to be tightly controlled by monoallelic expression. IL-2 can act as a growth hormone for both B and T lymphocytes. The human gene for interleukin 2(IL2) is assigned to chromosome 4.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat IL-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each rat IL-2 standard solution and each sample is measured in duplicate.