Human BMP-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0150
76236-104EA 1110.74 CAD
76236-104
Human BMP-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human BMP-2 AccuSignal ELISA Kit
  • Natural and recombinant human BMP-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard:CHO ; Immunogen sequence: Q283-R396
Bone morphogenetic protein-2(BMP-2, BMP 2A) belongs to the transforming growth factor beta(TGF-beta) superfamily. It is thought to be involved in cartilage and bone formation during embryogenesis, but may have additional functions in morphogenesis as implied by its expression in various organs and embryonic tissues of mice.1 BMP-2 has been identified as a candidate mediator of retinoid activity. BMP-2 protein induces medulloblastoma cell apoptosis, whereas the BMP-2 antagonist noggin blocks both retinoid and BMP-2-induced apoptosis. BMP-2 also induces p38 mitogen-activated protein kinase(MAPK), which is necessary for BMP-2 and retinoid-induced apoptosis.2 Bone morphogenic proteins(BMPs) are known to promote osteogenesis, and clinical trials are currently underway to evaluate the ability of certain BMPs to promote fracture-healing and spinal fusion.3 The standard product used in this kit is recombinant human BMP-2, constituting dimer by two chains of 114 amino acids with the molecular mass of 26KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human BMP-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human bone tissue, cell culture supernates or serum to each empty well. It is recommended that each human BMP-2 standard solution and each sample be measured in duplicate.