Anti-IL1RA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-401-303-0100
76235-122EA 834.24 CAD
76235-122
Anti-IL1RA Rabbit Polyclonal Antibody
Antibodies
Interleukin 1 receptor antagonist is secreted by various types of cells including immune, epithelial and adipocytes. IL 1Ra is used to treat rheumatoid arthritis. Anti-IL-1 RA antibody is ideal for investigators involved in Cancer and Immunology research.

This IgG fraction antibody of anti-Human IL-1 Receptor Antagonist has been tested for use in neutralizations, ELISA,  radioimmunoassays, western blotting, immunohistochemistry and flow cytometry.  Reactivity in other immunoassays is unknown.This product has been assayed by immunoblot using HRP Goat-anti-Rabbit IgG [H&L] (More Product Information
Order Now

SPECIFICATIONS

Antigen symbol IL1RA
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Purification DEAE fractionation
Storage temperature Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR