Anti-MAP2K2 Mouse Monoclonal Antibody (DL800) [Clone: 19G10.F1.E2]
Supplier: Rockland Immunochemical
200-345-GN8S
76235-058EA 224.52 CAD
76235-058
Anti-MAP2K2 Mouse Monoclonal Antibody (DL800) [Clone: 19G10.F1.E2]
Antibodies
MEK2 antibodies detect the MEK2 isoform.  Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation.   This pathway also plays a key role in synaptic plasticity in the brain.  Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase.   MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain.   The MEK2 antibody is ideal for investigators involved inMore Product Information