Anti-IKBKG Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemical
200-401-GM7S
76234-936EA 229.1 CAD
76234-936
Anti-IKBKG Rabbit Polyclonal Antibody
Antibodies
Anti-NEMO antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Anti-NEMO antibody detects recombinant and endogenous NEMO. NEMO, the regulatory subunit of the IKK core complex, phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. Its binding to scaffolding polyubiquitin seems to play a role in IKK activation by multiple signaling receptor pathways. Nemo is also considered to be a mediator for TAX activation of NF-kappa-B and may beMore Product Information