Anti-MAP2K2 Mouse Monoclonal Antibody (HRP) [Clone: 19G10.F1.E2]
Supplier: Rockland Immunochemical
200-303-GN8
76234-916EA 840.34 CAD
76234-916
Anti-MAP2K2 Mouse Monoclonal Antibody (HRP) [Clone: 19G10.F1.E2]
Antibodies
MEK2 antibodies detect the MEK2 isoform. Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling andMore Product Information