Anti-MAP2K2 Mouse Monoclonal Antibody [Clone: 19G10.F1.E2]

Supplier: Rockland Immunochemical
200-301-GN8S
76234-906EA 229.1 CAD
76234-906
Anti-MAP2K2 Mouse Monoclonal Antibody [Clone: 19G10.F1.E2]
Antibodies
MEK2 antibodies detect the MEK2 isoform.  Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation.   This pathway also plays a key role in synaptic plasticity in the brain.  Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase.   MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain.   The MEK2 antibody is ideal for investigators involved inMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol MAP2K2
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Clone 19G10.F1.E2
Reactivity
Host Mouse
Isotype IgG2b kappa
Western blot Yes
ELISA Yes
Purification Protein A purified
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR