Anti-IL8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
109-401-311-0100
76234-812EA 834.24 CAD
76234-812
Anti-IL8 Rabbit Polyclonal Antibody
Antibodies
Interleukin-8 is a chemokine produced by macrophages and other cell types such as epithelial cells. Human IL-8 (CXCL8) antibody is commonly used in many experiments focused around inflammation. Anti-IL-8 antibody is ideal for investigators involved in Cancer, Immunology and Cardiovascular research.

IL 8 antibody has been tested for use in ELISA and recognizes IL-8 found in cells, supernatants and other body fluids.  Reactivity in other immunoassays is unknown.

Type: Primary
Antigen: IL8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR