Anti-IgG Rabbit Mouse Polyclonal Antibody (DL680)

Supplier: Rockland Immunochemical
610-444-C46
76225-992EA 216.46 CAD
76225-992
Anti-IgG Rabbit Mouse Polyclonal Antibody (DL680)
Antibodies
Anti-Mouse IgG DyLight680 Antibody generated in rabbit detects reactivity to Mouse IgG1, IgG2a, IgG2b and IgG3. Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(More Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation DyLight® 680
Reactivity
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Purification immunoaffinity chromatography
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR