Llama IgG (FITC (Fluorescein Isothiocyanate))
Supplier: Rockland Immunochemical
025-0202
76221-912EA 304.7 CAD
76221-912
Llama IgG (FITC (Fluorescein Isothiocyanate))
Antibodies
Llama IgG Fluorescein Conjugate
  • control or standard reagent
  • multiplex, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting
Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. Such antibodies are homodimers of heavy chains that lack the CH1 domain of conventional antibodies and thereforeMore Product Information