Llama IgG2 isotype control (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemical
025-0244
76221-894EA 304.7 CAD
76221-894
Llama IgG2 isotype control (FITC (Fluorescein Isothiocyanate))
Antibodies
Llama IgG2 isotype control Fluorescein Conjugate

  • control or standard reagent
  • multiplex, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting

Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. Such antibodies are homodimers of heavy chains that lack the CH1 domain of conventionalMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG2
Environmentally Preferable
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity
Host
Isotype
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR