4-Amino-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Aminopyrazole, 1H-Pyrazol-4-amine

P2523-1G
76220-454EA 123.24 CAD
76220-454
4-Amino-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Amino-1H-pyrazole
Formula: C₃H₅N₃
MDL Number: MFCD01693729
CAS Number: 28466-26-4

Order Now

Specification Test Results

Appearance White to Brown to Dark purple powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR